missing translation for 'onlineSavingsMsg'
Learn More

3-Aminooxetane-3-carboxylic acid, 95%

Catalog No. AAH57291MD
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57291MD 250 mg
AAH57291ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH57291MD Supplier Thermo Scientific Chemicals Supplier No. H57291MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 138650-24-5 | C4H7NO3 | 117.104 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 138650-24-5
Molecular Formula C4H7NO3
Molecular Weight (g/mol) 117.104
MDL Number MFCD11035888
InChI Key LXRNYRAHXDUNKR-UHFFFAOYSA-N
Synonym 3-oxetanecarboxylic acid, 3-amino, 3-amino-oxetane-3-carboxylic acid, acmc-20a7wi, 3-amino-3-oxetanecarboxylic acid, 3-aminooxetane-3-carboxylic acid
PubChem CID 15024256
IUPAC Name 3-aminooxetane-3-carboxylic acid
SMILES C1C(CO1)(C(=O)O)N

Specifications

Melting Point 205°C to 210°C
Color White
Quantity 250 mg
Formula Weight 117.1
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 3-Aminooxetane-3-carboxylic acid
Hazard Category H302-H319
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only