missing translation for 'onlineSavingsMsg'
Learn More

3-Aminomethyltetrahydrofuran, 97%

Catalog No. p-4229065
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433000010 1 g Glass Bottle
AC433000050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC433000010 Supplier Thermo Scientific Chemicals Supplier No. 433000010

CAS: 165253-31-6 | C5H11NO | 101.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 165253-31-6
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD08234925
InChI Key CINJIXGRSTYIHP-UHFFFAOYSA-N
Synonym tetrahydrofuran-3-yl methanamine, tetrahydro-3-furanmethanamine, tetrahydrofuran-3-ylmethylamine, 3-furanmethanamine, tetrahydro, 1-tetrahydrofuran-3-ylmethanamine, c-tetrahydro-furan-3-yl-methylamine, 3-aminomethyl oxolane, 3-aminomethyl tetrahydrofuran, tetrahydrofur-3-yl methylamine, c-tetrahydrofuran-3-yl methylamine
PubChem CID 10898660
IUPAC Name oxolan-3-ylmethanamine
SMILES C1COCC1CN

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Boiling Point 65.0°C (15.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
UN Number 2924
Formula Weight 101.15
Percent Purity 97%
Physical Form Solution
Chemical Name or Material 3-Aminomethyltetrahydrofuran
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

RUO – Research Use Only