missing translation for 'onlineSavingsMsg'
Learn More

3-(Aminomethyl)-1-tert-butoxycarbonylpiperidine 98.0+%, TCI America™

Catalog No. A28931G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28931G 1 g
A28935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28931G Supplier TCI America Supplier No. A28931G

Chemical Identifiers

CAS 162167-97-7
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD01317792
InChI Key WPWXYQIMXTUMJB-UHFFFAOYSA-N
Synonym tert-butyl 3-aminomethyl piperidine-1-carboxylate, 1-boc-3-aminomethylpiperidine, 1-boc-3-aminomethyl piperidine, 3-aminomethyl-1-n-boc-piperidine, 1-boc-3-aminomethyl-piperidine, 3-aminomethyl-1-boc-piperidine, 3-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester, n-boc-3-aminomethylpiperidine, 1-n-boc-3-aminomethyl-piperidine
PubChem CID 2756483
IUPAC Name tert-butyl 3-(aminomethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)CN

Specifications

Color Yellow
Quantity 1 g
Formula Weight 214.31
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 3-(Aminomethyl)-1-tert-butoxycarbonylpiperidine