missing translation for 'onlineSavingsMsg'
Learn More

3-Aminomethyl-1-Boc-piperidine, 97%

Catalog No. AAH5283206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5283206 5 g
AAH5283203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5283206 Supplier Thermo Scientific Chemicals Supplier No. H5283206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 162167-97-7 | C11H22N2O2 | 214.309 g/mol

3-Aminomethyl-1-Boc-piperidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminomethyl-1-Boc-piperidine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 162167-97-7
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD01317792
InChI Key WPWXYQIMXTUMJB-UHFFFAOYSA-N
Synonym tert-butyl 3-aminomethyl piperidine-1-carboxylate, 1-boc-3-aminomethylpiperidine, 1-boc-3-aminomethyl piperidine, 3-aminomethyl-1-n-boc-piperidine, 1-boc-3-aminomethyl-piperidine, 3-aminomethyl-1-boc-piperidine, 3-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester, n-boc-3-aminomethylpiperidine, 1-n-boc-3-aminomethyl-piperidine
PubChem CID 2756483
IUPAC Name tert-butyl 3-(aminomethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)CN

Specifications

Quantity 5 g
Formula Weight 214.31
Percent Purity 97%
Chemical Name or Material 3-Aminomethyl-1-Boc-piperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only