missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminoisonicotinic acid, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 7579-20-6 |
|---|---|
| Molecular Formula | C6H5N2O2 |
| Molecular Weight (g/mol) | 137.12 |
| MDL Number | MFCD00137840 |
| InChI Key | FYEQKMAVRYRMBL-UHFFFAOYSA-M |
| Synonym | 3-aminoisonicotinic acid, 3-amino-4-pyridinecarboxylic acid, 3-amino-isonicotinic acid, 3-amino-4-pyridine carboxylic acid, 4-pyridinecarboxylic acid, 3-amino, 3-amino-4-carboxypyridine, 3-amino-4-pyridinecarboxylicacid, 3-amino-4-carboxylic acid pyridine, pubchem2420, 3-aminoisonicotinsaure |
| PubChem CID | 459531 |
| IUPAC Name | 3-aminopyridine-4-carboxylic acid |
| SMILES | NC1=CN=CC=C1C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC457690050
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| C6H5N2O2 | |
| MFCD00137840 | |
| 3-aminoisonicotinic acid, 3-amino-4-pyridinecarboxylic acid, 3-amino-isonicotinic acid, 3-amino-4-pyridine carboxylic acid, 4-pyridinecarboxylic acid, 3-amino, 3-amino-4-carboxypyridine, 3-amino-4-pyridinecarboxylicacid, 3-amino-4-carboxylic acid pyridine, pubchem2420, 3-aminoisonicotinsaure | |
| 3-aminopyridine-4-carboxylic acid |
Specifications
| 7579-20-6 | |
| 97.5% min. (HPLC) | |
| MFCD00137840 | |
| FYEQKMAVRYRMBL-UHFFFAOYSA-M | |
| 3-aminopyridine-4-carboxylic acid | |
| 459531 | |
| 98% |
| Authentic | |
| C6H5N2O2 | |
| 3-aminoisonicotinic acid, 3-amino-4-pyridinecarboxylic acid, 3-amino-isonicotinic acid, 3-amino-4-pyridine carboxylic acid, 4-pyridinecarboxylic acid, 3-amino, 3-amino-4-carboxypyridine, 3-amino-4-pyridinecarboxylicacid, 3-amino-4-carboxylic acid pyridine, pubchem2420, 3-aminoisonicotinsaure | |
| NC1=CN=CC=C1C([O-])=O | |
| 137.12 | |
| 138.13 | |
| 3-Aminoisonicotinic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
Warning
RUO – Research Use Only