missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminofluoranthene 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 120°C |
| Color | Yellow |
| Quantity | 5 g |
| UN Number | 3077 |
| Formula Weight | 217.27 |
| Percent Purity | ≥96.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Aminofluoranthene |
Safety and Handling
| RTECSNumber | LL4000000 |
| TSCA | No |