missing translation for 'onlineSavingsMsg'
Learn More

3-Aminofluoranthene 96.0+%, TCI America™

Catalog No. A27891G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27891G 1 g
A27895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27891G Supplier TCI America Supplier No. A27891G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2693-46-1
Molecular Formula C16H11N
Molecular Weight (g/mol) 217.271
MDL Number MFCD00001185
InChI Key VHGJAFIHUSTRGB-UHFFFAOYSA-N
Synonym 3-aminofluoranthene, 3-fluoranthenamine, 4-aminofluoranthene, fluoranthen-3-ylamine, 4-fluoranthenamine, unii-8un5prh090, ccris 7011, fluoranthene-3-ylamine, 3-aminofluoroanthene, timtec-bb sbb003376
PubChem CID 17599
IUPAC Name fluoranthen-3-amine
SMILES C1=CC=C2C(=C1)C3=C4C2=CC=CC4=C(C=C3)N

Specifications

Melting Point 120°C
Color Yellow
Quantity 1 g
UN Number 3077
Formula Weight 217.27
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminofluoranthene
RTECSNumber LL4000000
TSCA No