missing translation for 'onlineSavingsMsg'
Learn More

3-Aminocyclohexanecarboxylic Acid (cis- and trans- mixture) 95.0+%, TCI America™

Catalog No. p-7117043
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A07495G 5 g
A074925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A07495G Supplier TCI America Supplier No. A07495G

Chemical Identifiers

CAS 25912-50-9
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD00059562
InChI Key CKTUXQBZPWBFDX-UHFFFAOYNA-N
PubChem CID 544887
IUPAC Name 3-aminocyclohexane-1-carboxylic acid
SMILES NC1CCCC(C1)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 143.19
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminocyclohexanecarboxylic Acid (cis- and trans- mixture)