missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminocyclohexanecarboxylic Acid (cis- and trans- mixture) 95.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Formula Weight | 143.19 |
| Percent Purity | ≥95.0% (T) |
| Grade | Research |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Aminocyclohexanecarboxylic Acid (cis- and trans- mixture) |
Safety and Handling
| TSCA | No |