missing translation for 'onlineSavingsMsg'
Learn More

3-Aminocrotononitrile 98.0+%, TCI America™

Catalog No. p-7117145
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A094425G 25 g
A0944100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A094425G Supplier TCI America Supplier No. A094425G
Only null left

Chemical Identifiers

CAS 1118-61-2
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00008071,MFCD00008071
InChI Key DELJOESCKJGFML-DUXPYHPUSA-N
Synonym 3-aminocrotononitrile, 2z-3-aminobut-2-enenitrile, 3-amino-2-butenenitrile, z-3-aminobut-2-enenitrile, beta-aminocrotononitrile, 3-amino-2-butenonitrile, 2-amino-1-propenecarbonitrile, 3-amino-crotononitrile, beta-amino-crotononitrile, 2z-3-amino-2-butenenitrile
PubChem CID 5325263
IUPAC Name (2E)-3-aminobut-2-enenitrile
SMILES C\C(N)=C/C#N

Specifications

Melting Point 75°C
Color Yellow
Boiling Point 120°C
Quantity 25 g
Formula Weight 82.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminocrotononitrile
EINECSNumber (2)-1526
TSCA Yes