missing translation for 'onlineSavingsMsg'
Learn More

3-Aminocrotononitrile, (E)+(Z), 96%

Catalog No. AAL0531622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0531622 100 g
AAL0531636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0531622 Supplier Thermo Scientific Chemicals Supplier No. L0531622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1118-61-2 | C4H6N2 | 82.11 g/mol

3-Aminocrotononitrile reacts with ferrocenyl-1,2-enones to form ferrocenyl pyridines. It undergoes diazotization coupling reaction with p-substituted anilines to give 2-arylhydrazone-3-ketimino-butyronitriles.3-Aminocrotononitrile, (E)+(Z), is used in reaction with aryldiazonium salts gave, instead of the expected triazene, 2-arylhydrazono-3-ketobutyronitrile by electrophilic attack at the ?-carbon and hydrolysis of the resulting imine. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 3-Aminocrotononitrile was used as bisnucleophilic reagent which on cyclocondensation with hexafluoroacetone(ethoxycarbonylimine) forms bis(trifluoromethyl)pyrimidinones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminocrotononitrile reacts with ferrocenyl-1,2-enones to form ferrocenyl pyridines. It undergoes diazotization coupling reaction with p-substituted anilines to give 2-arylhydrazone-3-ketimino-butyronitriles.3-Aminocrotononitrile, (E)+(Z), is used in reaction with aryldiazonium salts gave, instead of the expected triazene, 2-arylhydrazono-3-ketobutyronitrile by electrophilic attack at the ɑ-carbon and hydrolysis of the resulting imine. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. 3-Aminocrotononitrile was used as bisnucleophilic reagent which on cyclocondensation with hexafluoroacetone(ethoxycarbonylimine) forms bis(trifluoromethyl)pyrimidinones.

Solubility
Very soluble in water, soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 1118-61-2
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00008071,MFCD00008071
InChI Key DELJOESCKJGFML-DUXPYHPUSA-N
Synonym 3-aminocrotononitrile, 2z-3-aminobut-2-enenitrile, 3-amino-2-butenenitrile, z-3-aminobut-2-enenitrile, beta-aminocrotononitrile, 3-amino-2-butenonitrile, 2-amino-1-propenecarbonitrile, 3-amino-crotononitrile, beta-amino-crotononitrile, 2z-3-amino-2-butenenitrile
PubChem CID 5325263
IUPAC Name (Z)-3-aminobut-2-enenitrile
SMILES C\C(N)=C/C#N

Specifications

Melting Point ∼70°C to 75°C
Boiling Point 120°C (4 mmHg)
Flash Point 154°C (309°F)
Quantity 100 g
Beilstein 1719815
Solubility Information Very soluble in water,soluble in ethanol.
Formula Weight 82.11
Percent Purity 96%
Chemical Name or Material 3-Aminocrotononitrile, (E)+(Z)
Hazard Category H302+H332-H317
Precautionary Statement P261-P264b-P270-P271-P272-P280g-P301+P312-P302+P352-P304+P340-P312-P330-P333+P313-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 214-266-2
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only