missing translation for 'onlineSavingsMsg'
Learn More

3-Aminocoumarin 98.0+%, TCI America™

Catalog No. A2816200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2816200MG 200 mg
A28161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2816200MG Supplier TCI America Supplier No. A2816200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1635-31-0
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00016965
InChI Key QWZHDKGQKYEBKK-UHFFFAOYSA-N
Synonym 3-aminocoumarin, 3-amino-2h-chromen-2-one, 3-amino-2-benzopyrone, 3-amino-2h-1-benzopyran-2-one, coumarin amine, coumarin monoamine, amino-chromen-2-one, 3-azanylchromen-2-one, acmc-1bous
PubChem CID 74217
IUPAC Name 3-aminochromen-2-one
SMILES C1=CC=C2C(=C1)C=C(C(=O)O2)N

Specifications

Melting Point 137°C
Color Yellow
Quantity 200 mg
Formula Weight 161.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminocoumarin