missing translation for 'onlineSavingsMsg'
Learn More

3-Aminocoumarin, 97%

Catalog No. AAL14031ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14031ME 500 mg
AAL14031MC 100 mg
AAL1403104 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL14031ME Supplier Thermo Scientific Chemicals Supplier No. L14031ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1635-31-0 | C9H7NO2 | 161.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1635-31-0
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00016965
InChI Key QWZHDKGQKYEBKK-UHFFFAOYSA-N
Synonym 3-aminocoumarin, 3-amino-2h-chromen-2-one, 3-amino-2-benzopyrone, 3-amino-2h-1-benzopyran-2-one, coumarin amine, coumarin monoamine, amino-chromen-2-one, 3-azanylchromen-2-one, acmc-1bous
PubChem CID 74217
IUPAC Name 3-aminochromen-2-one
SMILES C1=CC=C2C(=C1)C=C(C(=O)O2)N

Specifications

Melting Point 135°C to 137°C
Quantity 500 mg
Beilstein 128033
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 3-Aminocoumarin
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-659-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only