missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzofuran-2-carboxamide 98.0+%, TCI America™

Catalog No. A28325G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28325G 5 g
A28321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28325G Supplier TCI America Supplier No. A28325G
Only null left

Chemical Identifiers

CAS 54802-10-7
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.18
MDL Number MFCD00457147
InChI Key FXOYSSLDDDZIJN-UHFFFAOYSA-N
PubChem CID 600546
IUPAC Name 3-amino-1-benzofuran-2-carboxamide
SMILES NC(=O)C1=C(N)C2=CC=CC=C2O1

Specifications

Melting Point 159°C
Color White-Yellow
Quantity 5 g
Formula Weight 176.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminobenzofuran-2-carboxamide