missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzenesulfonic Acid 99.0+%, TCI America™

Catalog No. p-7116796
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A026525G 25 g
A0265500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A026525G Supplier TCI America Supplier No. A026525G

Chemical Identifiers

CAS 121-47-1
Molecular Formula C6H7NO3S
Molecular Weight (g/mol) 173.186
MDL Number MFCD00065345
InChI Key ZAJAQTYSTDTMCU-UHFFFAOYSA-N
Synonym metanilic acid, m-sulfanilic acid, m-aminobenzenesulfonic acid, m-anilinesulfonic acid, 1-aminobenzene-3-sulfonic acid, benzenesulfonic acid, 3-amino, aminobenzenesulfonic acid, kyselina metanilova, 3-amino-benzenesulfonic acid, 3-sulfoaniline
PubChem CID 8474
ChEBI CHEBI:27764
IUPAC Name 3-aminobenzenesulfonic acid
SMILES C1=CC(=CC(=C1)S(=O)(=O)O)N

Specifications

Quantity 25 g
Formula Weight 173.19
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminobenzenesulfonic Acid
EINECSNumber (3)-1971
RTECSNumber OY2300000
TSCA Yes