missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzeneboronic acid hydrochloride, 98%

Catalog No. p-7034895
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5259503 1 g
AAH5259506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5259503 Supplier Thermo Scientific Chemicals Supplier No. H5259503
Only null left

CAS: 85006-23-1 | C6H9BClNO2 | 173.40 g/mol

3-Aminobenzeneboronic acid hydrochloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminobenzeneboronic acid hydrochloride is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 85006-23-1
Molecular Formula C6H9BClNO2
Molecular Weight (g/mol) 173.40
MDL Number MFCD00191748
InChI Key QBMHZZHJIBUPOX-UHFFFAOYSA-N
Synonym 3-aminophenyl boronic acid hydrochloride, 3-aminophenylboronic acid hydrochloride, 3-aminophenylboronic acid hcl, m-aminophenylboronic acid hydrochloride, 3-aminophenylboronic acid, hcl, 3-aminobenzeneboronic acid hydrochloride, boronic acid, 3-aminophenyl-, hydrochloride, pubchem1751, pubchem20396, timtec-bb sbb003816
PubChem CID 2763241
IUPAC Name (3-aminophenyl)boronic acid;hydrochloride
SMILES Cl.NC1=CC=CC(=C1)B(O)O

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 173.41
Percent Purity 98%
Chemical Name or Material 3-Aminobenzeneboronic acid hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 285-052-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only