missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-N-(tert-butoxycarbonyl)benzylamine 98.0+%, TCI America™

Catalog No. p-7118038
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27181G 1 g
A27185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27181G Supplier TCI America Supplier No. A27181G
Only null left

Chemical Identifiers

CAS 147291-66-5
Molecular Formula C12H18N2O2
Molecular Weight (g/mol) 222.288
MDL Number MFCD04974010
InChI Key LSOZALWRNWQPLK-UHFFFAOYSA-N
Synonym 3-Amino-N-Boc-benzylamine, 3-[(tert-Butoxycarbonylamino)methyl]aniline, 3-[(Boc-amino)methyl]aniline, alpha-(tert-Butoxycarbonyl)amino-m-toluidine, alpha-(Boc-amino)-m-toluidine, tert-Butyl (3-Aminobenzyl)carbamate, (3-Aminobenzyl)carbamic Acid tert-Butyl
PubChem CID 10775412
IUPAC Name tert-butyl N-[(3-aminophenyl)methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1=CC(=CC=C1)N

Specifications

Melting Point 50°C
Color White-Yellow
Quantity 1 g
Formula Weight 222.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-N-(tert-butoxycarbonyl)benzylamine