missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-9H-pyrido[3,4-b]indole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 291°C |
| Quantity | 1 g |
| Formula Weight | 183.21 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Amino-9H-pyrido[3,4-b]indole |
Safety and Handling
| RTECSNumber | UU9351650 |
| TSCA | No |