missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-6-morpholinopyridazine, 97%

Catalog No. AC433270050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC433270050 5 g
AC433270010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC433270050 Supplier Thermo Scientific Chemicals Supplier No. 433270050

CAS: 66346-91-6 | C8H12N4O | 180.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66346-91-6
Molecular Formula C8H12N4O
Molecular Weight (g/mol) 180.21
InChI Key IEUHTZVAUDMKQJ-UHFFFAOYSA-N
Synonym 6-morpholinopyridazin-3-amine, 3-amino-6-morpholin-4-yl pyridazine, 6-morpholin-4-yl pyridazin-3-amine, 3-amino-6-morpholin-4-ylpyridazine, 6-morpholin-4-yl-pyridazin-3-ylamine, 3-pyridazinamine, 6-4-morpholinyl, 6-morpholinopyridazine-3-amine, 6-4-morpholinyl-3-pyridazinamine, 3-amino-6-morpholin-4-3-amino-6-morpholin-4-ylpyridazine
PubChem CID 13114295
IUPAC Name 6-morpholin-4-ylpyridazin-3-amine
SMILES C1COCCN1C2=NN=C(C=C2)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 181°C to 183°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 180.21
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 3-Amino-6-morpholinopyridazine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only