missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-6-chloropyridazine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 5469-69-2
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.55
MDL Number MFCD00051506
InChI Key DTXVKPOKPFWSFF-UHFFFAOYSA-N
Synonym 3-amino-6-chloropyridazine, 3-pyridazinamine, 6-chloro, 6-chloro-3-pyridazinamine, 6-amino-3-chloropyridazine, 6-chloro-3-aminopyridazine, 6-chloropyridazin-3-ylamine, 6-chloro-pyridazin-3-ylamine, unii-ijs443slzg, 3-chloro-6-pyridazinamine, 6-chloro-3-pyridazinylamine
PubChem CID 21643
IUPAC Name 6-chloropyridazin-3-amine
SMILES NC1=CC=C(Cl)N=N1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC382830500
SDS View Documents
Thermo Scientific Chemicals
382830500
50g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5469-69-2
129.55
DTXVKPOKPFWSFF-UHFFFAOYSA-N
21643
NC1=CC=C(Cl)N=N1
C4H4ClN3
MFCD00051506
3-amino-6-chloropyridazine, 3-pyridazinamine, 6-chloro, 6-chloro-3-pyridazinamine, 6-amino-3-chloropyridazine, 6-chloro-3-aminopyridazine, 6-chloropyridazin-3-ylamine, 6-chloro-pyridazin-3-ylamine, unii-ijs443slzg, 3-chloro-6-pyridazinamine, 6-chloro-3-pyridazinylamine
6-chloropyridazin-3-amine

Specifications

5469-69-2
Glass bottle
MFCD00051506
3-amino-6-chloropyridazine, 3-pyridazinamine, 6-chloro, 6-chloro-3-pyridazinamine, 6-amino-3-chloropyridazine, 6-chloro-3-aminopyridazine, 6-chloropyridazin-3-ylamine, 6-chloro-pyridazin-3-ylamine, unii-ijs443slzg, 3-chloro-6-pyridazinamine, 6-chloro-3-pyridazinylamine
NC1=CC=C(Cl)N=N1
129.55
129.55
3-Amino-6-chloropyridazine
97.5% min. (HPLC)
C4H4ClN3
50g
DTXVKPOKPFWSFF-UHFFFAOYSA-N
6-chloropyridazin-3-amine
21643
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

EINECSNumber : 226-796-1

SDS
Documents

Documents

RUO – Research Use Only