missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-6-chloro-2-methylpyridine, 95%, Thermo Scientific Chemicals

Catalog No. AAH6146506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6146506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6146506 Supplier Thermo Scientific Chemicals Supplier No. H6146506

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 164666-68-6
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.586
MDL Number MFCD03095220
InChI Key QVCIIOZINFCMDJ-UHFFFAOYSA-N
Synonym 3-amino-6-chloro-2-picoline, 3-amino-6-chloro-2-methylpyridine, 5-amino-2-chloro-6-methylpyridine, 5-amino-2-chloro-6-picoline, 6-chloro-2-methyl-pyridin-3-ylamine, 3-pyridinamine, 6-chloro-2-methyl, 6-chloro-2-methyl-3-pyridylamine, 6-chloro-2-methyl-3-pyridinamine, 2-chloro-5-amino-6-methylpyridine, pubchem1246
PubChem CID 7009500
IUPAC Name 6-chloro-2-methylpyridin-3-amine
SMILES CC1=C(C=CC(=N1)Cl)N

Specifications

Color Yellow
Quantity 5 g
Formula Weight 142.59
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Amino-6-chloro-2-methylpyridine
Hazard Category H315-H319
Hazard Statement GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only