missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-6-bromo-2-chloropyridine 98.0+%, TCI America™

Catalog No. A29675G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29675G 5 g
A29671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29675G Supplier TCI America Supplier No. A29675G
Only null left

Chemical Identifiers

CAS 169833-70-9
Molecular Formula C5H4BrClN2
Molecular Weight (g/mol) 207.455
MDL Number MFCD04971930
InChI Key YCECOMBEVDPUJG-UHFFFAOYSA-N
PubChem CID 2050085
IUPAC Name 6-bromo-2-chloropyridin-3-amine
SMILES C1=CC(=NC(=C1N)Cl)Br

Specifications

Melting Point 119°C
Quantity 5 g
Formula Weight 207.46
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-6-bromo-2-chloropyridine