missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-phenylpyrazole 98.0+%, TCI America™

Catalog No. A21021G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21021G 1 g
A21025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21021G Supplier TCI America Supplier No. A21021G

Chemical Identifiers

CAS 1572-10-7
Molecular Formula C9H9N3
Molecular Weight (g/mol) 159.192
MDL Number MFCD00191749
InChI Key PWSZRRFDVPMZGM-UHFFFAOYSA-N
Synonym 3-phenyl-1h-pyrazol-5-amine, 3-amino-5-phenylpyrazole, 5-amino-3-phenylpyrazole, 1h-pyrazol-3-amine, 5-phenyl, 5-phenyl-2h-pyrazol-3-amine, pyrazole, 5-amino-3-phenyl, 3-amino-5-phenyl-1h-pyrazole, 5-phenyl-2h-pyrazol-3-ylamine, 1h-pyrazol-5-amine, 3-phenyl, 5-phenyl-1h-pyrazol-3-ylamine
PubChem CID 136655
IUPAC Name 5-phenyl-1H-pyrazol-3-amine
SMILES C1=CC=C(C=C1)C2=CC(=NN2)N

Specifications

Melting Point 126°C
Quantity 1 g
Formula Weight 159.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-5-phenylpyrazole
RTECSNumber UQ6109000
TSCA No