missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-phenyl-1,2,4-thiadiazole, 96%

Catalog No. p-7032181
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33552MD 250 mg
AAH3355203 1 g
AAH3355206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH33552MD Supplier Thermo Scientific Chemicals Supplier No. H33552MD
Only null left

CAS: 27182-54-3 | C8H7N3S | 177.225 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27182-54-3
Molecular Formula C8H7N3S
Molecular Weight (g/mol) 177.225
MDL Number MFCD00159799
InChI Key WOZDQVLZCXNJPY-UHFFFAOYSA-N
Synonym 3-amino-5-phenyl-1,2,4-thiadiazole, 1,2,4-thiadiazol-3-amine, 5-phenyl, 5-phenyl-1,2,4-thiadiazol-3-yl-amine, 5-phenyl-1,2,4-thiadiazole-3-amine, #
PubChem CID 554203
IUPAC Name 5-phenyl-1,2,4-thiadiazol-3-amine
SMILES C1=CC=C(C=C1)C2=NC(=NS2)N

Specifications

Melting Point 136°C to 137°C
Quantity 250 mg
Formula Weight 177.23
Percent Purity 96%
Chemical Name or Material 3-Amino-5-phenyl-1,2,4-thiadiazole
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only