missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-mercapto-1,2,4-triazole, 97+%

Catalog No. p-7022454
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1524206 5 g
AAA1524214 25 g
AAA1524222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1524206 Supplier Thermo Scientific Chemicals Supplier No. A1524206
Only null left

CAS: 16691-43-3 | C2H4N4S | 116.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16691-43-3
Molecular Formula C2H4N4S
Molecular Weight (g/mol) 116.14
MDL Number MFCD00005231
InChI Key WZUUZPAYWFIBDF-UHFFFAOYSA-N
Synonym 3-amino-5-mercapto-1,2,4-triazole, iminothiourazole, 3-imino-5-thiourazole, 3-amino-1h-1,2,4-triazole-5-thiol, 5-amino-4h-1,2,4-triazole-3-thiol, 3-amino-1,2,4-triazole-5-thiol, s-triazole-3-thiol, 5-amino, 3h-1,2,4-triazole-3-thione, 5-amino-1,2-dihydro, 5-amino-1,2-dihydro-3h-1,2,4-triazole-3-thione, unii-353f2d8l1z
PubChem CID 2723869
SMILES NC1=NC(=S)NN1

Specifications

Melting Point >300°C (decomposition)
Odor Odorless
Quantity 5 g
Beilstein 112404
IUPAC Name 5-amino-1,2-dihydro-1,2,4-triazole-3-thione
Formula Weight 116.15
Percent Purity ≥98%
Chemical Name or Material 3-Amino-5-mercapto-1,2,4-triazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 240-735-6
RTECSNumber XZ5357500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only