missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-bromobenzotrifluoride, 97%, Thermo Scientific™

Chemical Identifiers

CAS 54962-75-3
Molecular Formula C7H5BrF3N
Molecular Weight (g/mol) 240.02
InChI Key HJTLKVYOWNTDPF-UHFFFAOYSA-N
Synonym 3-amino-5-bromobenzotrifluoride, 3-bromo-5-trifluoromethyl aniline, 3-bromo-5-trifluoromethyl-phenylamine, 3-amino-5-bromo trifluoromethyl benzene, 3-bromo-5-trifluoromethyl benzenamine, 3-bromo-5-aminobenzotrifluoride, 3-amino-5-bromotrifluorotoluene, benzenamine, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluoromethyl phenylamine
PubChem CID 2735880
IUPAC Name 3-bromo-5-(trifluoromethyl)aniline
SMILES C1=C(C=C(C=C1N)Br)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC442660050
SDS View Documents
Thermo Scientific Chemicals
442660050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC442660250
SDS View Documents
Thermo Scientific Chemicals
442660250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

54962-75-3
240.02
3-amino-5-bromobenzotrifluoride, 3-bromo-5-trifluoromethyl aniline, 3-bromo-5-trifluoromethyl-phenylamine, 3-amino-5-bromo trifluoromethyl benzene, 3-bromo-5-trifluoromethyl benzenamine, 3-bromo-5-aminobenzotrifluoride, 3-amino-5-bromotrifluorotoluene, benzenamine, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluoromethyl phenylamine
3-bromo-5-(trifluoromethyl)aniline
C7H5BrF3N
HJTLKVYOWNTDPF-UHFFFAOYSA-N
2735880
C1=C(C=C(C=C1N)Br)C(F)(F)F

Specifications

54962-75-3
100.0
220.0°C to 223.0°C
96% min. (GC)
5g
3-amino-5-bromobenzotrifluoride, 3-bromo-5-trifluoromethyl aniline, 3-bromo-5-trifluoromethyl-phenylamine, 3-amino-5-bromo trifluoromethyl benzene, 3-bromo-5-trifluoromethyl benzenamine, 3-bromo-5-aminobenzotrifluoride, 3-amino-5-bromotrifluorotoluene, benzenamine, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluoromethyl phenylamine
C1=C(C=C(C=C1N)Br)C(F)(F)F
240.02
240.02
3-Amino-5-bromobenzotrifluoride
96.0
1.6970g/mL
Authentic
C7H5BrF3N
1.697
HJTLKVYOWNTDPF-UHFFFAOYSA-N
3-bromo-5-(trifluoromethyl)aniline
2735880
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 259-412-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only