missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-bromo-2-methoxypyridine, 96%, Thermo Scientific Chemicals

Catalog No. p-7036056
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5429906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5429906 Supplier Thermo Scientific Chemicals Supplier No. H5429906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 884495-39-0 | Molecular Formula: C6H7BrN2O | Molecular Weight: 203.039 g/mol

3-Amino-5-bromo-2-methoxypyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-5-bromo-2-methoxypyridine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 884495-39-0
Molecular Formula C6H7BrN2O
Molecular Weight (g/mol) 203.039
MDL Number MFCD07368887
InChI Key HJOOFLFWIISCAI-UHFFFAOYSA-N
Synonym 3-amino-5-bromo-2-methoxypyridine, 5-bromo-2-methoxy-3-pyridinamine, 3-amino-5-bromopyridin-2-yl methyl ether, 5-bromo-2-methoxy-3-aminopyridine, 5-bromo-2-methoxy-3-cyanopyridine, 5-bromo-2-methyloxy-3-pyridinamine, 3-pyridinamine,5-bromo-2-methoxy, 5-bromo-2-methoxy-pyridin-3-ylamine, pubchem17164, acmc-209qtg
PubChem CID 44754869
IUPAC Name 5-bromo-2-methoxypyridin-3-amine
SMILES COC1=NC=C(C=C1N)Br

Specifications

Melting Point 53°C to 55°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 203.04
Percent Purity 96%
Chemical Name or Material 3-Amino-5-bromo-2-methoxypyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only