missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-(4-chlorophenyl)pyrazole 98.0+%, TCI America™

Catalog No. p-7118166
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
A29521G 1 g
A29525G 5 g
2 options

Catalog No. A29521G

Supplier: TCI America A29521G

Only null left

Chemical Identifiers

CAS 78583-81-0
Molecular Formula C9H8ClN3
Molecular Weight (g/mol) 193.63
MDL Number MFCD00053046
InChI Key XQPBZIITFQHIDI-UHFFFAOYSA-N
Synonym 3-4-chlorophenyl-1h-pyrazol-5-amine, 5-4-chlorophenyl-1h-pyrazol-3-amine, 5-amino-3-4-chlorophenyl pyrazole, 5-4-chlorophenyl-2h-pyrazol-3-amine, 3-amino-5-4-chlorophenyl-1h-pyrazole, 5-amino-3-4-chlorophenyl-1h-pyrazole, 3-amino-5-4-chlorophenyl pyrazole, 5-4-chlorophenyl-2h-pyrazol-3-ylamine, 3-4-chlorophenyl pyrazole-5-ylamine, pubchem12126
PubChem CID 2735305
IUPAC Name 5-(4-chlorophenyl)-1H-pyrazol-3-amine
SMILES NC1=NNC(=C1)C1=CC=C(Cl)C=C1

Specifications

Melting Point 175°C
Color White
Quantity 1 g
Formula Weight 193.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-5-(4-chlorophenyl)pyrazole