missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-(trifluoromethyl)pyridine 98.0+%, TCI America™

Catalog No. A29715G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29715G 5 g
A29711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29715G Supplier TCI America Supplier No. A29715G

Chemical Identifiers

CAS 175204-80-5
Molecular Formula C6H5F3N2
Molecular Weight (g/mol) 162.12
MDL Number MFCD00173960
InChI Key DROFUWWORPKKSI-UHFFFAOYSA-N
Synonym 3-amino-4-trifluoromethyl pyridine, 4-trifluoromethyl pyridin-3-amine, 3-amino-4-trifluoromethylpyridine, 4-trifluoromethyl-pyridin-3-ylamine, 3-pyridinamine, 4-trifluoromethyl, 4-trifluoromethyl-3-pyridinamine, 4-trifluoromethyl-3-pyridylamine, pubchem2987, acmc-209e9a, 4-trifluoromethyl-3-pyridinylamine
PubChem CID 2735957
IUPAC Name 4-(trifluoromethyl)pyridin-3-amine
SMILES NC1=C(C=CN=C1)C(F)(F)F

Specifications

Melting Point 56°C
Quantity 5 g
Formula Weight 162.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Amino-4-(trifluoromethyl)pyridine