missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-4-methylpyridine 98.0+%, TCI America™
$94.33 - $277.23
Chemical Identifiers
| CAS | 3430-27-1 |
|---|---|
| Molecular Formula | C6H8N2 |
| Molecular Weight (g/mol) | 108.144 |
| MDL Number | MFCD00128871 |
| InChI Key | IBKMZYWDWWIWEL-UHFFFAOYSA-N |
| Synonym | 3-amino-4-methylpyridine, 3-amino-4-picoline, 3-pyridinamine, 4-methyl, 4-methyl-3-pyridinamine, 4-methyl-pyridin-3-ylamine, unii-9hn55rq07b, 4-methyl-3-pyridylamine, 4-methylpyridine-3-amine, 3-amino-4-methyl pyridine, 3-amino-4-methylpyridine 3-amino-4-picoline |
| PubChem CID | 137935 |
| IUPAC Name | 4-methylpyridin-3-amine |
| SMILES | CC1=C(C=NC=C1)N |
Chemical Identifiers
| 3430-27-1 | |
| 108.144 | |
| IBKMZYWDWWIWEL-UHFFFAOYSA-N | |
| 137935 | |
| CC1=C(C=NC=C1)N |
| C6H8N2 | |
| MFCD00128871 | |
| 3-amino-4-methylpyridine, 3-amino-4-picoline, 3-pyridinamine, 4-methyl, 4-methyl-3-pyridinamine, 4-methyl-pyridin-3-ylamine, unii-9hn55rq07b, 4-methyl-3-pyridylamine, 4-methylpyridine-3-amine, 3-amino-4-methyl pyridine, 3-amino-4-methylpyridine 3-amino-4-picoline | |
| 4-methylpyridin-3-amine |
Specifications
| 3430-27-1 | |
| 254°C | |
| MFCD00128871 | |
| 2811 | |
| IBKMZYWDWWIWEL-UHFFFAOYSA-N | |
| 4-methylpyridin-3-amine | |
| 137935 | |
| ≥98.0% (GC,T) | |
| 3-Amino-4-methylpyridine |
| 106°C | |
| C6H8N2 | |
| 5 g | |
| 3-amino-4-methylpyridine, 3-amino-4-picoline, 3-pyridinamine, 4-methyl, 4-methyl-3-pyridinamine, 4-methyl-pyridin-3-ylamine, unii-9hn55rq07b, 4-methyl-3-pyridylamine, 4-methylpyridine-3-amine, 3-amino-4-methyl pyridine, 3-amino-4-methylpyridine 3-amino-4-picoline | |
| CC1=C(C=NC=C1)N | |
| 108.144 | |
| 108.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No