missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-ethyl-1H-pyrazole, 98%

Catalog No. p-7054788
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6319903 1 g
AAH6319906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6319903 Supplier Thermo Scientific Chemicals Supplier No. H6319903

CAS: 43024-15-3 | C5H9N3 | 111.15 g/mol

3-Amino-4-ethyl-1H-pyrazole is employed as a intermediate for chemical research and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-4-ethyl-1H-pyrazole is employed as a intermediate for chemical research and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 43024-15-3
Molecular Formula C5H9N3
Molecular Weight (g/mol) 111.15
MDL Number MFCD06797570
InChI Key RDCODVKTTJWFAR-UHFFFAOYSA-N
Synonym 4-ethyl-1h-pyrazol-3-amine, 3-amino-4-ethylpyrazole, 3-amino-4-ethyl-1h-pyrazole, 4-ethyl-2h-pyrazol-3-amine, 3-amino-4-ethyl pyrazole, 4-ethyl-1h-pyrazol-3-ylamine, 1h-pyrazol-3-amine, 4-ethyl
PubChem CID 11789157
IUPAC Name 4-ethyl-1H-pyrazol-5-amine
SMILES CCC1=C(N)NN=C1

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 111.15
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Amino-4-ethyl-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only