missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-4-chloropyridine 98.0+%, TCI America™
$78.29 - $233.00
Chemical Identifiers
| CAS | 20511-15-3 |
|---|---|
| Molecular Formula | C5H5ClN2 |
| Molecular Weight (g/mol) | 128.559 |
| MDL Number | MFCD00235124 |
| InChI Key | GTLFLMZOABSJSV-UHFFFAOYSA-N |
| Synonym | 3-amino-4-chloropyridine, 4-chloro-pyridin-3-ylamine, 4-chlorpyridin-3-amin, 4-chloro-3-pyridinamine, 4-chloro-3-aminopyridine, 3-pyridinamine, 4-chloro, 4-chloro-3-pyridylamine, 4-chloropyridin-3-ylamine, pubchem6665, acmc-209fbc |
| PubChem CID | 581932 |
| IUPAC Name | 4-chloropyridin-3-amine |
| SMILES | C1=CN=CC(=C1Cl)N |
Chemical Identifiers
| 20511-15-3 | |
| 128.559 | |
| GTLFLMZOABSJSV-UHFFFAOYSA-N | |
| 581932 | |
| C1=CN=CC(=C1Cl)N |
| C5H5ClN2 | |
| MFCD00235124 | |
| 3-amino-4-chloropyridine, 4-chloro-pyridin-3-ylamine, 4-chlorpyridin-3-amin, 4-chloro-3-pyridinamine, 4-chloro-3-aminopyridine, 3-pyridinamine, 4-chloro, 4-chloro-3-pyridylamine, 4-chloropyridin-3-ylamine, pubchem6665, acmc-209fbc | |
| 4-chloropyridin-3-amine |
Specifications
| 20511-15-3 | |
| C5H5ClN2 | |
| 1 g | |
| GTLFLMZOABSJSV-UHFFFAOYSA-N | |
| 4-chloropyridin-3-amine | |
| 581932 | |
| ≥98.0% (GC,T) | |
| 3-Amino-4-chloropyridine |
| 60°C | |
| MFCD00235124 | |
| 3-amino-4-chloropyridine, 4-chloro-pyridin-3-ylamine, 4-chlorpyridin-3-amin, 4-chloro-3-pyridinamine, 4-chloro-3-aminopyridine, 3-pyridinamine, 4-chloro, 4-chloro-3-pyridylamine, 4-chloropyridin-3-ylamine, pubchem6665, acmc-209fbc | |
| C1=CN=CC(=C1Cl)N | |
| 128.559 | |
| 128.56 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator