missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-4-chlorobenzonitrile 97.0+%, TCI America™
Supplier: TCI America A21051G
Specifications
| 3-Amino-4-chlorobenzonitrile | |
| 53312-79-1 | |
| Crystal-Powder at 20°C | |
| MFCD00017089 | |
| 2-Chloro-5-cyanoaniline | |
| NC1=CC(=CC=C1Cl)C#N | |
| 152.58 | |
| 152.58 |
| 90°C | |
| 1 g | |
| C7H5ClN2 | |
| 3439 | |
| KRBRYJLBILJHAS-UHFFFAOYSA-N | |
| 3-amino-4-chlorobenzonitrile | |
| 104464 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 53312-79-1 | |
| 152.58 | |
| KRBRYJLBILJHAS-UHFFFAOYSA-N | |
| 104464 | |
| NC1=CC(=CC=C1Cl)C#N |
| C7H5ClN2 | |
| MFCD00017089 | |
| 2-Chloro-5-cyanoaniline | |
| 3-amino-4-chlorobenzonitrile |
Safety and Handling
EINECSNumber : (3)-1817
TSCA : No