missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-chloro-1H-indazole, 97%

Catalog No. AAH34143MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34143MD 250 mg
AAH3414303 1 g
AAH3414306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH34143MD Supplier Thermo Scientific Chemicals Supplier No. H34143MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20925-60-4 | C7H6ClN3 | 167.596 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20925-60-4
Molecular Formula C7H6ClN3
Molecular Weight (g/mol) 167.596
MDL Number MFCD00663139
InChI Key QPLXQNVPEHUPTR-UHFFFAOYSA-N
Synonym 3-amino-4-chloro-1h-indazole, 1h-indazol-3-amine, 4-chloro, 3-amino-4-chloroindazole, 4-chloro-1h-indazole-3-amine, 4-chloro-1h-indazol-3-ylamine, 3-amino4-chloroindazole, 4-chloro-1h-indazole-3-ylamine
PubChem CID 313444
IUPAC Name 4-chloro-1H-indazol-3-amine
SMILES C1=CC2=C(C(=C1)Cl)C(=NN2)N

Specifications

Melting Point 157°C to 159°C
Quantity 250 mg
Formula Weight 167.6
Percent Purity 97%
Chemical Name or Material 3-Amino-4-chloro-1H-indazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only