missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-carbethoxypyrazole, 99%

Catalog No. AC103310050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC103310050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC103310050 Supplier Thermo Scientific Chemicals Supplier No. 103310050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6994-25-8 | C6H9N3O2 | 155.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

CAS 6994-25-8
CAS Min % 98.5
CAS Max % 100.0
Melting Point 103.0°C to 107.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C6H9N3O2
Quantity 5 g
MDL Number MFCD00005238
Synonym ethyl 3-amino-1h-pyrazole-4-carboxylate, ethyl 3-amino-4-pyrazolecarboxylate, 3-amino-4-carbethoxypyrazole, 5-amino-1h-pyrazole-4-carboxylic acid ethyl ester, 3-amino-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl 3-aminopyrazole-4-carboxylate, 3-amino-4-ethoxycarbonylpyrazole, unii-9gy5ha1gaa, 3-amino-4-carbethoxypyrazol, 1h-pyrazole-4-carboxylic acid, 3-amino-, ethyl ester
InChI Key YPXGHKWOJXQLQU-UHFFFAOYSA-N
SMILES CCOC(=O)C1=C(NN=C1)N
IUPAC Name ethyl 5-amino-1H-pyrazole-4-carboxylate
Molecular Weight (g/mol) 155.16
PubChem CID 81472
Formula Weight 155.16
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-carbethoxypyrazole, 99%
Show More Show Less
EINECSNumber 230-262-3

RUO – Research Use Only