missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-bromopyrazole 98.0+%, TCI America™

$67.38 - $265.67

Chemical Identifiers

CAS 16461-94-2
Molecular Formula C3H4BrN3
Molecular Weight (g/mol) 161.99
MDL Number MFCD00082728
InChI Key OELYMZVJDKSMOJ-UHFFFAOYSA-N
Synonym 3-amino-4-bromopyrazole, 4-bromo-1h-pyrazol-3-amine, 5-amino-4-bromo-1h-pyrazole, 3-amino-4-bromo-1h-pyrazole, 1h-pyrazol-3-amine, 4-bromo, 4-bromo-2h-pyrazol-3-amine, 4-bromo-1h-pyrazol-3-ylamine, 4-bromo-2h-pyrazol-3-ylamine, 4-bromopyrazole-5-ylamine, 3-amino-4-bromo pound inverted question markpyrazole
PubChem CID 140079
IUPAC Name 4-bromo-1H-pyrazol-5-amine
SMILES C1=NNC(=C1Br)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A26881G
View Documents
TCI America
A26881G
1 g
Each for $67.38
Only null left
 
A26885G
View Documents
TCI America
A26885G
5 g
Each for $265.67
Only null left
 
Description

Description

Specifications

Chemical Identifiers

16461-94-2
161.99
OELYMZVJDKSMOJ-UHFFFAOYSA-N
140079
C1=NNC(=C1Br)N
C3H4BrN3
MFCD00082728
3-amino-4-bromopyrazole, 4-bromo-1h-pyrazol-3-amine, 5-amino-4-bromo-1h-pyrazole, 3-amino-4-bromo-1h-pyrazole, 1h-pyrazol-3-amine, 4-bromo, 4-bromo-2h-pyrazol-3-amine, 4-bromo-1h-pyrazol-3-ylamine, 4-bromo-2h-pyrazol-3-ylamine, 4-bromopyrazole-5-ylamine, 3-amino-4-bromo pound inverted question markpyrazole
4-bromo-1H-pyrazol-5-amine

Specifications

16461-94-2
White-Yellow
MFCD00082728
3-amino-4-bromopyrazole, 4-bromo-1h-pyrazol-3-amine, 5-amino-4-bromo-1h-pyrazole, 3-amino-4-bromo-1h-pyrazole, 1h-pyrazol-3-amine, 4-bromo, 4-bromo-2h-pyrazol-3-amine, 4-bromo-1h-pyrazol-3-ylamine, 4-bromo-2h-pyrazol-3-ylamine, 4-bromopyrazole-5-ylamine, 3-amino-4-bromo pound inverted question markpyrazole
C1=NNC(=C1Br)N
161.99
161.99
Crystalline Powder
138°C
C3H4BrN3
1 g
OELYMZVJDKSMOJ-UHFFFAOYSA-N
4-bromo-1H-pyrazol-5-amine
140079
≥98.0% (GC,T)
3-Amino-4-bromopyrazole
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents