missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-bromo-5-methylisoxazole, 97%

Catalog No. AAB2013803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2013803 1 g
AAB2013806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2013803 Supplier Thermo Scientific Chemicals Supplier No. B2013803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5819-40-9 | C4H5BrN2O | 177.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5819-40-9
Molecular Formula C4H5BrN2O
Molecular Weight (g/mol) 177.00
MDL Number MFCD00052553
InChI Key JEZOZNWEHSNXPQ-UHFFFAOYSA-N
Synonym 3-amino-4-bromo-5-methylisoxazole, 4-bromo-5-methylisoxazol-3-amine, 3-isoxazolamine, 4-bromo-5-methyl, 4-bromo-5-methylisoxazole-3-ylamine, acmc-20ana7, 4-bromo-5-methyl-3-isoxazolamine, 4-bromo-5-methyl-isoxazol-3-ylamine, 4-bromanyl-5-methyl-1,2-oxazol-3-amine, 3-amino-4-bromo-5-methyl isoxazole
PubChem CID 2774464
IUPAC Name 4-bromo-5-methyl-1,2-oxazol-3-amine
SMILES CC1=C(Br)C(N)=NO1

Specifications

Melting Point 73°C to 75°C
Quantity 1 g
Formula Weight 177.02
Percent Purity 97%
Chemical Name or Material 3-Amino-4-bromo-5-methylisoxazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only