missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acide 3-Amino-3-Phénylpropionique 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 223°C |
| Color | Blanc |
| Quantity | 5 g |
| Formula Weight | 165.19 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | 3-Amino-3-phenylpropionic Acid |
Sécurité et manipulation
| TSCA | No |