missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-3-phenylpropionic acid, 99%
CAS: 614-19-7 | C9H11NO2 | 165.19 g/mol
Chemical Identifiers
| CAS | 614-19-7 |
|---|---|
| Molecular Formula | C9H11NO2 |
| Molecular Weight (g/mol) | 165.19 |
| MDL Number | MFCD00008064,MFCD01076238 |
| InChI Key | UJOYFRCOTPUKAK-UHFFFAOYNA-N |
| Synonym | 3-amino-3-phenylpropionic acid, dl-3-amino-3-phenylpropionic acid, dl-beta-phenylalanine, beta-phenyl-beta-alanine, 3-phenyl-dl-beta-alanine, h-dl-beta-phe-oh, benzenepropanoic acid, beta-amino, 3-amino-3-phenyl-propionic acid, 3a3ppa, beta-aminohydrocinnamic acid |
| PubChem CID | 69189 |
| ChEBI | CHEBI:68528 |
| IUPAC Name | 3-amino-3-phenylpropanoic acid |
| SMILES | NC(CC(O)=O)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAL1980103
|
N/A | 1 g |
N/A
|
|
|||||
|
AAL1980106
|
N/A | 5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C9H11NO2 | |
| MFCD00008064,MFCD01076238 | |
| 3-amino-3-phenylpropionic acid, dl-3-amino-3-phenylpropionic acid, dl-beta-phenylalanine, beta-phenyl-beta-alanine, 3-phenyl-dl-beta-alanine, h-dl-beta-phe-oh, benzenepropanoic acid, beta-amino, 3-amino-3-phenyl-propionic acid, 3a3ppa, beta-aminohydrocinnamic acid | |
| CHEBI:68528 | |
| NC(CC(O)=O)C1=CC=CC=C1 |
Specifications
| ∽220°C (decomposition) | |
| C9H11NO2 | |
| 2803286 | |
| UJOYFRCOTPUKAK-UHFFFAOYNA-N | |
| 3-amino-3-phenylpropanoic acid | |
| 69189 | |
| 165.19 | |
| 3-Amino-3-phenylpropionic acid |
| 614-19-7 | |
| MFCD00008064,MFCD01076238 | |
| 3-amino-3-phenylpropionic acid, dl-3-amino-3-phenylpropionic acid, dl-beta-phenylalanine, beta-phenyl-beta-alanine, 3-phenyl-dl-beta-alanine, h-dl-beta-phe-oh, benzenepropanoic acid, beta-amino, 3-amino-3-phenyl-propionic acid, 3a3ppa, beta-aminohydrocinnamic acid | |
| NC(CC(O)=O)C1=CC=CC=C1 | |
| 165.19 | |
| CHEBI:68528 | |
| 99% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-371-2
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only