missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-3-phenylpropionic acid, 99%

Catalog No. aal1980106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1980106 5 g
AAL1980103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1980106 Supplier Thermo Scientific Chemicals Supplier No. L1980106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 614-19-7 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 614-19-7
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00008064,MFCD01076238
InChI Key UJOYFRCOTPUKAK-UHFFFAOYNA-N
Synonym 3-amino-3-phenylpropionic acid, dl-3-amino-3-phenylpropionic acid, dl-beta-phenylalanine, beta-phenyl-beta-alanine, 3-phenyl-dl-beta-alanine, h-dl-beta-phe-oh, benzenepropanoic acid, beta-amino, 3-amino-3-phenyl-propionic acid, 3a3ppa, beta-aminohydrocinnamic acid
PubChem CID 69189
ChEBI CHEBI:68528
IUPAC Name 3-amino-3-phenylpropanoic acid
SMILES NC(CC(O)=O)C1=CC=CC=C1

Specifications

Melting Point ∽220°C (decomposition)
Beilstein 2803286
Quantity 5 g
Formula Weight 165.19
Percent Purity 99%
Chemical Name or Material 3-Amino-3-phenylpropionic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-371-2
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only