missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-3-phenyl-1-propanol 97.0+%, TCI America™
$42.87 - $117.43
Chemical Identifiers
| CAS | 14593-04-5 |
|---|---|
| Molecular Formula | C9H13NO |
| Molecular Weight (g/mol) | 151.21 |
| MDL Number | MFCD00801124,MFCD01311768,MFCD01311791 |
| InChI Key | SEQXIQNPMQTBGN-UHFFFAOYNA-N |
| Synonym | 3-amino-3-phenyl-1-propanol, dl-beta-phenylalaninol, 3-amino-3-phenyl-propan-1-ol, 3-phenyl-dl-beta-alaninol, 1-propanol, 3-amino-3-phenyl, benzenepropanol, gamma-amino, 1-amino-3-hydroxyprop-1-yl benzene, dl--phenylalaninol, 3-amino-3-phenylpropanol, acmc-1c4c0 |
| PubChem CID | 203500 |
| IUPAC Name | 3-amino-3-phenylpropan-1-ol |
| SMILES | NC(CCO)C1=CC=CC=C1 |
Chemical Identifiers
| 14593-04-5 | |
| 151.21 | |
| SEQXIQNPMQTBGN-UHFFFAOYNA-N | |
| 203500 | |
| NC(CCO)C1=CC=CC=C1 |
| C9H13NO | |
| MFCD00801124,MFCD01311768,MFCD01311791 | |
| 3-amino-3-phenyl-1-propanol, dl-beta-phenylalaninol, 3-amino-3-phenyl-propan-1-ol, 3-phenyl-dl-beta-alaninol, 1-propanol, 3-amino-3-phenyl, benzenepropanol, gamma-amino, 1-amino-3-hydroxyprop-1-yl benzene, dl--phenylalaninol, 3-amino-3-phenylpropanol, acmc-1c4c0 | |
| 3-amino-3-phenylpropan-1-ol |
Specifications
| 14593-04-5 | |
| White | |
| C9H13NO | |
| 1 g | |
| 3-amino-3-phenyl-1-propanol, dl-beta-phenylalaninol, 3-amino-3-phenyl-propan-1-ol, 3-phenyl-dl-beta-alaninol, 1-propanol, 3-amino-3-phenyl, benzenepropanol, gamma-amino, 1-amino-3-hydroxyprop-1-yl benzene, dl--phenylalaninol, 3-amino-3-phenylpropanol, acmc-1c4c0 | |
| NC(CCO)C1=CC=CC=C1 | |
| 151.21 | |
| 151.21 | |
| Crystalline Powder |
| 75°C | |
| 134°C | |
| MFCD00801124,MFCD01311768,MFCD01311791 | |
| 3259 | |
| SEQXIQNPMQTBGN-UHFFFAOYNA-N | |
| 3-amino-3-phenylpropan-1-ol | |
| 203500 | |
| ≥97.0% (GC,T) | |
| 3-Amino-3-phenyl-1-propanol |
Safety and Handling
RTECSNumber : UA6903000
TSCA : No