missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-3-methyloxetane 97.0+%, TCI America™

Catalog No. p-7118078
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2792200MG 200 mg
A27921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2792200MG Supplier TCI America Supplier No. A2792200MG
Only null left

Chemical Identifiers

CAS 874473-14-0
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.122
MDL Number MFCD11111713
InChI Key NQVWMPOQWBDSAI-UHFFFAOYSA-N
Synonym 3-methyl-3-oxetanamine, 3-amino-3-methyloxetane, 3-oxetanamine,3-methyl, 3-oxetanamine, 3-methyl, pubchem14416, pubchem23160, acmc-209qnl, 3-methyl 3-oxetanamine, 3-methyl-3-oxetaneamine, 3-amino-3-methyl-oxetane
PubChem CID 46835725
IUPAC Name 3-methyloxetan-3-amine
SMILES CC1(COC1)N

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 87.12
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Amino-3-methyloxetane