missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-3-(4-methoxyphenyl)propionic Acid 98.0+%, TCI America™

Catalog No. A25325G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25325G 5 g
A25321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25325G Supplier TCI America Supplier No. A25325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5678-45-5
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.218
MDL Number MFCD00187209
InChI Key NYTANCDDCQVQHG-UHFFFAOYSA-N
Synonym 3-amino-3-4-methoxyphenyl propanoic acid, 3-amino-3-4-methoxyphenyl propionic acid, 3-amino-3-4-methoxy-phenyl-propionic acid, 3-p-methoxyphenyl-dl-beta-alanine, dl-3-amino-3-4-methoxy-phenyl-propionic acid, dl-beta-p-methoxyphenyl alanine, beta-4-methoxyphenyl-beta-aminopropionic acid, beta-amino-4-methoxybenzenepropanoic acid, rs-beta-amino-beta-4-methoxyphenyl propionic acid, s-3-amino-3-4-methoxyphenyl-propionic acid
PubChem CID 585889
IUPAC Name 3-amino-3-(4-methoxyphenyl)propanoic acid
SMILES COC1=CC=C(C=C1)C(CC(=O)O)N

Specifications

Melting Point 232°C
Color White
Quantity 5 g
Formula Weight 195.22
Percent Purity ≥98.0% (HPLC,T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-3-(4-methoxyphenyl)propionic Acid