missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-3-(3,4-dimethoxyphenyl)propionic Acid 98.0+%, TCI America™

Catalog No. A25365G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25365G 5 g
A25361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25365G Supplier TCI America Supplier No. A25365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34841-09-3
Molecular Formula C11H15NO4
Molecular Weight (g/mol) 225.244
MDL Number MFCD00735172
InChI Key FGCXSFRGPCUBPW-UHFFFAOYSA-N
Synonym 3-amino-3-3,4-dimethoxyphenyl propanoic acid, 3-amino-3-3,4-dimethoxyphenyl propionic acid, 3-amino-3-3,4-dimethoxy-phenyl-propionic acid, dl-beta-3,4-dimethoxyphenyl alanine, 3-3,4-dimethoxyphenyl-dl-beta-alanine, dl-3-amino-3-3,4-dimethoxy-phenyl-propionic acid, 3-3,4-dimethoxyphenyl-beta-alanine, 3-amino-3-3,4-dimethoxy-phenyl propionic acid, 3-amino-3-3,4-dimethoxyphenyl propionoic acid, pubchem14766
PubChem CID 597182
IUPAC Name 3-amino-3-(3,4-dimethoxyphenyl)propanoic acid
SMILES COC1=C(C=C(C=C1)C(CC(=O)O)N)OC

Specifications

Melting Point 228°C
Color White
Quantity 5 g
Formula Weight 225.24
Percent Purity ≥98.0% (HPLC,T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-3-(3,4-dimethoxyphenyl)propionic Acid