missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-methylpyridine 98.0+%, TCI America™

Catalog No. A23335G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A23335G Supplier TCI America Supplier No. A23335G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory reagent used in synthetic applications.

Chemical Identifiers

CAS 3430-10-2
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD03788195
InChI Key ZSFPJJJRNUZCEV-UHFFFAOYSA-N
Synonym 3-amino-2-picoline, 3-amino-2-methylpyridine, 2-methyl-3-pyridinamine, 2-methyl-3-aminopyridine, 2-methyl-pyridin-3-ylamine, 3-pyridinamine, 2-methyl, 2-methyl-3-pyridinylamine, 2-methyl-3-pyridylamine, 2-methylpyridine-3-amine, zlchem 29
PubChem CID 572616
IUPAC Name 2-methylpyridin-3-amine
SMILES CC1=C(C=CC=N1)N

Specifications

Melting Point 116°C
Quantity 5 g
UN Number 2811
Formula Weight 108.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2-methylpyridine