missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-2-methyl-6-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 106°C to 108°C |
| Quantity | 1 g |
| Synonym | 2-methyl-6-trifluoromethyl pyridin-3-amine, 2-methyl-6-trifluoromethyl-3-pyridinamine, 3-amino-2-methyl-6-trifluoromethyl pyridine, 2-methyl-6-trifluoromethylpyridin-3-yl amine, 2-methyl-6-trifluoromethyl-pyridin-3-ylamine, 2-methyl-6-trifluoromethyl-3-pyridylamine, 3-pyridinamine,2-methyl-6-trifluoromethyl, 2-methyl-6-trifluoromethyl pyridine-3-amine, 3-pyridinamine, 2-methyl-6-trifluoromethyl |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 176.14 |
| Percent Purity | 97% |
| Chemical Name or Material | 3-Amino-2-methyl-6-(trifluoromethyl)pyridine |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |