missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-cyclohexen-1-one, 95%, Thermo Scientific™

Catalog No. p-4812899
Change view
Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC219310050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC219310050 Supplier Thermo Scientific Chemicals Supplier No. 219310050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5220-49-5
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.14
MDL Number MFCD00013783
InChI Key ZZMRPOAHZITKBV-UHFFFAOYSA-N
Synonym 3-amino-2-cyclohexen-1-one, 3-aminocyclohex-2-enone, 2-cyclohexen-1-one, 3-amino, unii-v596547zke, 3-amino-2-cyclohexen-1-ketone, 3-amino-2-cyclohexenone, 3-aminocyclohexenone, 3-aminocyclohex-enone, pubchem14449, acmc-209kyd
PubChem CID 78892
IUPAC Name 3-aminocyclohex-2-en-1-one
SMILES NC1=CC(=O)CCC1

Specifications

Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5g
Formula Weight 111.14
Percent Purity 95%
Chemical Name or Material 3-Amino-2-cyclohexen-1-one
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 226-014-9

RUO – Research Use Only