missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-2-bromopyridine 98.0+%, TCI America™
$61.51 - $190.13
Chemical Identifiers
| CAS | 39856-58-1 |
|---|---|
| Molecular Formula | C5H5BrN2 |
| Molecular Weight (g/mol) | 173.013 |
| InChI Key | HKDVVTLISGIPFE-UHFFFAOYSA-N |
| Synonym | 3-amino-2-bromopyridine, 2-bromo-3-aminopyridine, 2-bromo-pyridineamine, 3-pyridinamine, 2-bromo, 2-bromo-3-pyridinamine, 2-bromo-3-pyridylamine, 2-bromopyridine-3-amine, 3-amino-2-bromo-pyridine, 2-bromo-3-pyridineamine, 2-bromo-pyridin-3-ylamine |
| PubChem CID | 642816 |
| IUPAC Name | 2-bromopyridin-3-amine |
| SMILES | C1=CC(=C(N=C1)Br)N |
Chemical Identifiers
| 39856-58-1 | |
| 173.013 | |
| 3-amino-2-bromopyridine, 2-bromo-3-aminopyridine, 2-bromo-pyridineamine, 3-pyridinamine, 2-bromo, 2-bromo-3-pyridinamine, 2-bromo-3-pyridylamine, 2-bromopyridine-3-amine, 3-amino-2-bromo-pyridine, 2-bromo-3-pyridineamine, 2-bromo-pyridin-3-ylamine | |
| 2-bromopyridin-3-amine |
| C5H5BrN2 | |
| HKDVVTLISGIPFE-UHFFFAOYSA-N | |
| 642816 | |
| C1=CC(=C(N=C1)Br)N |
Specifications
| 39856-58-1 | |
| C5H5BrN2 | |
| 3-amino-2-bromopyridine, 2-bromo-3-aminopyridine, 2-bromo-pyridineamine, 3-pyridinamine, 2-bromo, 2-bromo-3-pyridinamine, 2-bromo-3-pyridylamine, 2-bromopyridine-3-amine, 3-amino-2-bromo-pyridine, 2-bromo-3-pyridineamine, 2-bromo-pyridin-3-ylamine | |
| C1=CC(=C(N=C1)Br)N | |
| 173.013 | |
| 173.01 | |
| Crystalline Powder |
| 79°C | |
| 1 g | |
| HKDVVTLISGIPFE-UHFFFAOYSA-N | |
| 2-bromopyridin-3-amine | |
| 642816 | |
| ≥98.0% (GC,T) | |
| 3-Amino-2-bromopyridine |
Safety and Handling
TSCA : No