missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-bromopyridine, 97%

Catalog No. p-7047847
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1983903 1 g
AAL1983914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1983903 Supplier Thermo Scientific Chemicals Supplier No. L1983903

CAS: 39856-58-1 | C5H5BrN2 | 173.013 g/mol

Used in a palladium-catalyzed synthesis of indole systems containing fused medium- and large-ring heterocycles. pyrrole derivatives ahs been prepared using 3-Amino-2-bromopyridine. Used in P-C coupling reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in a palladium-catalyzed synthesis of indole systems containing fused medium- and large-ring heterocycles. pyrrole derivatives ahs been prepared using 3-Amino-2-bromopyridine. Used in P-C coupling reaction.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 39856-58-1
Molecular Formula C5H5BrN2
Molecular Weight (g/mol) 173.013
MDL Number MFCD00234064
InChI Key HKDVVTLISGIPFE-UHFFFAOYSA-N
Synonym 3-amino-2-bromopyridine, 2-bromo-3-aminopyridine, 2-bromo-pyridineamine, 3-pyridinamine, 2-bromo, 2-bromo-3-pyridinamine, 2-bromo-3-pyridylamine, 2-bromopyridine-3-amine, 3-amino-2-bromo-pyridine, 2-bromo-3-pyridineamine, 2-bromo-pyridin-3-ylamine
PubChem CID 642816
IUPAC Name 2-bromopyridin-3-amine
SMILES C1=CC(=C(N=C1)Br)N

Specifications

Melting Point 75°C to 79°C
Quantity 1 g
Beilstein 109830
Solubility Information Slightly soluble in water.
Formula Weight 173.01
Percent Purity 98%
Chemical Name or Material 3-Amino-2-bromopyridine
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only