missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2,6-dichloropyridine 98.0+%, TCI America™

Catalog No. p-7117529
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15805G 5 g
A158025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15805G Supplier TCI America Supplier No. A15805G
Only null left

Chemical Identifiers

CAS 62476-56-6
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.00
MDL Number MFCD00023417
InChI Key MJVZSRZTBDMYLX-UHFFFAOYSA-N
Synonym 3-amino-2,6-dichloropyridine, 2,6-dichloro-3-aminopyridine, 2,6-dichloro-3-pyridylamine, 3-pyridinamine, 2,6-dichloro, 2,6-dichloro-3-pyridinamine, 2,6-dichloropyridin-3-ylamine, 2,6-dichloro-pyridin-3-ylamine, 2,6-dichlorpyridin-3-amin, pubchem10384, ksc495k2f
PubChem CID 112846
IUPAC Name 2,6-dichloropyridin-3-amine
SMILES NC1=CC=C(Cl)N=C1Cl

Specifications

Melting Point 120°C
Quantity 5 g
Formula Weight 163.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2,6-dichloropyridine